Learn More
CTDP1/FCP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301171AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.Specifications
CTDP1/FCP1 | |
Polyclonal | |
Unconjugated | |
CTDP1 | |
CCFDN, FCP1, CTD of POLR2A, phosphatase of, subunit 1, RNA polymerase II subunit A C-terminal domain phosphatase, TFIIF-associating CTD phosphatase 1, serine phosphatase FCP1a, transcription factor IIF-associating CTD phosphatase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9150 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9Y5B0 | |
CTDP1 | |
Between 500 and 550 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.