Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTDSP1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP106029
Description
CTDSP1 Polyclonal specifically detects CTDSP1 in Human, Mouse, Rat, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
CTDSP1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 1, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 1, NIF3, NLI-IF, NLIIFEC 3.1.3.16, NLI-interacting factor 3, SCP1Nuclear LIM interactor-interacting factor 3, Small CTD phosphatase 1, Small C-terminal domain phosphatase 1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_067021.1and NP_872580.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:32000 | |
NP_067021.1 | |
CTDSP1 | |
Peptide with sequence C-RGKGDQKSAASQK corresponding to internal region according to NP_872580.1. | |
0.1 mg | |
Signal Transduction | |
58190 | |
Human, Mouse, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction