Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTDSP1 Antibody (OTI1B6), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP272110C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272110C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272110C Supplier Novus Biologicals Supplier No. NBP272110C
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTDSP1 Monoclonal specifically detects CTDSP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CTDSP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B6
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 1, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 1, NIF3, NLI-IF, NLIIFEC 3.1.3.16, NLI-interacting factor 3, SCP1Nuclear LIM interactor-interacting factor 3, Small CTD phosphatase 1, Small C-terminal domain phosphatase 1
Gene Symbols CTDSP1
Host Species Mouse
Immunogen Full length recombinant protein of human CTDSP1 (NP_067021) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 58190.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.