Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTDSP1 Monoclonal Antibody (OTI1C3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502266S


Description
CTDSP1 Monoclonal antibody specifically detects CTDSP1 in Human samples. It is validated for Western BlotSpecifications
CTDSP1 | |
Monoclonal | |
0.71 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96EY8 | |
Mouse | |
Affinity chromatography | |
RUO | |
326625 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI1C3 | |
Unconjugated | |
MMAB | |
carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 1; CTD; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 1; CTD small phosphatase 1; Ctdsp1; GIP; golli-interacting protein; Nif3; Nliif; NLI-IF; NLI-interacting factor 3; Nuclear LIM interactor-interacting factor 3; SCP1; small CTD phosphatase 1; Small C-terminal domain phosphatase 1 | |
Full length human recombinant protein of human MMAB produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction