Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTDSP2 Antibody (OTI7F5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271552F
Description
CTDSP2 Monoclonal specifically detects CTDSP2 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
CTDSP2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CTDSP2 | |
Full length recombinant protein of human CTDSP2 (NP_005721) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Immunohistochemistry | |
OTI7F5 | |
Immunohistochemistry | |
carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2, conserved gene amplified in osteosarcoma, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 2, NIF2, Nuclear LIM interactor-interacting factor 2, OS4NLI-interacting factor 2, Protein OS-4, PSR2, SCP2EC 3.1.3.16, Small CTD phosphatase 2, Small C-terminal domain phosphatase 2 | |
Mouse | |
Affinity Purified | |
Protein Phosphatase | |
10106.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction