Learn More
CTDSP2 Monoclonal Antibody (4F4)

Description
CTDSP2 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | CTDSP2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4F4 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CTDSP2 |
| Gene Accession No. | O14595 |
| Gene Alias | AI586070; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2; conserved gene amplified in osteosarcoma; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2; CTD small phosphatase 2; CTDSP2; D10Ertd73e; NIF2; NLI-interacting factor 2; nuclear LIM interactor-interacting factor 2; nuclear LIM interactor-interacting factor 2 (NLI-interacting factor 2) (Protein OS-4); nuclear LIM interactor-interacting factor 2-like; OS4; OS-4; protein OS-4; PSR2; SCP2; small CTD phosphatase 2; Small C-terminal domain phosphatase 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.