Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTDSP2 Monoclonal Antibody (OTI7F5)
GREENER_CHOICE

Catalog No. PIMA527044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527044 Supplier Invitrogen™ Supplier No. MA527044
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CTDSP2 Monoclonal Antibody for Western Blot, IHC (P)

CTDSP2 is one of several small CTD phosphatases (SCP). It has been proposed that SCP activity is an evolutionarily conserved transcriptional regulator that acts globally to silence neuronal genes. In addition, expression of phosphatase CTDSP2 mRNA isenhanced in several cancer cell lines with amplification of the CTDSP2 and CDK4 genes, and CTDSP2 coamplifies with CDK4 in some primary sarcomas. CTDSP2 may contribute to the development of a subset of sarcomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTDSP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CTDSP2
Gene Accession No. O14595
Gene Alias AI586070; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2; conserved gene amplified in osteosarcoma; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2; CTD small phosphatase 2; CTDSP2; D10Ertd73e; NIF2; NLI-interacting factor 2; nuclear LIM interactor-interacting factor 2; nuclear LIM interactor-interacting factor 2 (NLI-interacting factor 2) (Protein OS-4); nuclear LIM interactor-interacting factor 2-like; OS4; OS-4; protein OS-4; PSR2; SCP2; small CTD phosphatase 2; Small C-terminal domain phosphatase 2
Gene Symbols CTDSP2
Host Species Mouse
Immunogen Full length human recombinant protein of CTDSP2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10106
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.