Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTDSPL Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP120903
Description
CTDSPL Polyclonal specifically detects CTDSPL in Human, Canine samples. It is validated for Peptide ELISA.Specifications
CTDSPL | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
C3orf8, Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 3, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase-like, CTD small phosphatase-like protein, CTDSP-like, EC 3.1.3, HYA22, NIF1, NIFL, NIF-like protein, NLI-interacting factor 1, Nuclear LIM interactor-interacting factor 1, Protein YA22, PSR1, RB protein serine phosphatase from chromosome 3, RBSP3EC 3.1.3.16, SCP3chromosome 3 open reading frame 8, Small CTD phosphatase 3, Small C-terminal domain phosphatase 3, YA22 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both isoforms (NP_001008393.1; NP_005799.2). | |
Store at -20C. Avoid freeze-thaw cycles. |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:32000 | |
NP_001008393.1 | |
CTDSPL | |
Peptide with sequence QCNVSLKKQRSRS corresponding to internal region according to NP_001008393.1, NP_005799.2. | |
0.1 mg | |
Signal Transduction | |
10217 | |
Human, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction