Learn More
Invitrogen™ SCP3 Monoclonal Antibody (6F9C5)

Description
SCP3 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow, ELISA
Specifications
Specifications
| Antigen | SCP3 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6F9C5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% proprietary stabilizer and 0.05% sodium azide |
| Gene | Sycp3 |
| Gene Accession No. | Q8IZU3 |
| Gene Alias | Cor1; LOC615896 protein; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; Sycp3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.