Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTDSPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561713 Supplier Invitrogen™ Supplier No. PA561713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AIITQVTNPK EDEGRLPGAG EKASQCNVSL KKQRSRSILS SFFCCFRDYN VEAPPPSSPS VLPPLVEENG GLQKGDQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 41%.

Recruited by REST to neuronal genes that contain RE-1 elements, leading to neuronal gene silencing in non-neuronal cells. Preferentially catalyzes the dephosphorylation of 'Ser-5' within the tandem 7 residue repeats in the C-terminal domain (CTD) of the largest RNA polymerase II subunit POLR2A. Negatively regulates RNA polymerase II transcription, possibly by controlling the transition from initiation/capping to processive transcript elongation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTDSPL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CTDSPL
Gene Accession No. O15194
Gene Alias 2810418J22Rik; AI426263; C3orf8; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 3; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like; CTD small phosphatase like; CTD small phosphatase-like protein; CTDSPL; CTDSP-like; hYA22; MGC71888; Nif1; Nifl; NIF-like protein; NLI-interacting factor 1; nuclear LIM interactor-interacting factor 1; Protein YA22; PSR1; RB protein serine phosphatase from chromosome 3; RBSP3; SCP3; SCP-3; SCP3 COR1; small CTD phosphatase 3; Small C-terminal domain phosphatase 3; YA22
Gene Symbols CTDSPL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CTDSPL. Recombinant protein control fragment (Product #RP-100963).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10217
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.