Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTDSPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109305
Description
Positive Control: HepG2.
Recruited by REST to neuronal genes that contain RE-1 elements, leading to neuronal gene silencing in non-neuronal cells. Preferentially catalyzes the dephosphorylation of 'Ser-5' within the tandem 7 residue repeats in the C-terminal domain (CTD) of the largest RNA polymerase II subunit POLR2A. Negatively regulates RNA polymerase II transcription, possibly by controlling the transition from initiation/capping to processive transcript elongation. [UniProt].
Specifications
| CTDSPL | |
| Polyclonal | |
| Unconjugated | |
| CTDSPL | |
| 2810418J22Rik; AI426263; C3orf8; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 3; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like; CTD small phosphatase like; CTD small phosphatase-like protein; CTDSPL; CTDSP-like; hYA22; MGC71888; Nif1; Nifl; NIF-like protein; NLI-interacting factor 1; nuclear LIM interactor-interacting factor 1; Protein YA22; PSR1; RB protein serine phosphatase from chromosome 3; RBSP3; SCP3; SCP-3; SCP3 COR1; small CTD phosphatase 3; Small C-terminal domain phosphatase 3; YA22 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10217 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O15194 | |
| CTDSPL | |
| Recombinant protein within Human CTDSPL aa 77-252. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction