Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTDSPL Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988394
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8394 Supplier Bioss Supplier No. BS8241R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The synaptonemal complex is a proteinaceous complex that apparently mediates synapses during the zygotene stage and then disintegrates. It is a complex structure that unites homologous chromosomes during the prophase stage of meiosis.

Specifications

Antigen CTDSPL
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CTDSPL
Gene Accession No. O15194
Gene Alias 2810418J22Rik; AI426263; C3orf8; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 3; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like; CTD small phosphatase like; CTD small phosphatase-like protein; CTDSPL; CTDSP-like; hYA22; MGC71888; Nif1; Nifl; NIF-like protein; NLI-interacting factor 1; Nuclear LIM interactor-interacting factor 1; Protein YA22; PSR1; RB protein serine phosphatase from chromosome 3; RBSP3; SCP3; SCP-3; SCP3 COR1; small CTD phosphatase 3; Small C-terminal domain phosphatase 3; YA22
Gene Symbols CTDSPL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CTDSPL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10217
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.