Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTDSPL2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297397PE
Description
CTDSPL2 Polyclonal antibody specifically detects CTDSPL2 in Human samples. It is validated for Western BlotSpecifications
CTDSPL2 | |
Polyclonal | |
Western Blot | |
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase like 2, CTD small phosphatase-like protein 2, CTDSP-like 2, EC 3.1.3, EC 3.1.3.-, EC 3.1.3.16, FLJ10523, HSPC129 | |
Produced in rabbits immunized with E. coli-derived Human CTDSPL2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51496 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction