Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTDSPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105734
Description
Antibody detects endogenous levels of total CTDSPL2.
Probable phosphatase.
Specifications
| CTDSPL2 | |
| Polyclonal | |
| Unconjugated | |
| CTDSPL2 | |
| CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase like 2; CTD small phosphatase like 2; CTD small phosphatase-like protein 2; Ctdspl2; CTDSP-like 2; D2Ertd485e; HSPC058; HSPC129; RGD1309219 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 329506, 51496 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q05D32, Q8BG15 | |
| CTDSPL2 | |
| A synthesized peptide derived from human CTDSPL2(Accession Q05D32), corresponding to amino acid residues M1-L34. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction