Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTDSPL2 Polyclonal Antibody

Supplier: Thermo Scientific PA5115059
Description

Specifications
CTDSPL2 | |
Polyclonal | |
Unconjugated | |
CTDSPL2 | |
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase like 2; CTD small phosphatase like 2; CTD small phosphatase-like protein 2; Ctdspl2; CTDSP-like 2; D2Ertd485e; HSPC058; HSPC129; RGD1309219 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
311368, 329506, 51496 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q05D32, Q5XIK8, Q8BG15 | |
CTDSPL2 | |
A synthesized peptide derived from human CTDSPL2(Accession Q05D32), corresponding to amino acid residues T84-S134. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction