Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Ctf1 (Mouse) ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
Ctf1 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse CT-1 in cell culture supernates, serum and plasma (heparin, EDTA).
Specifications
Specifications
Product Type | ELISA Kit |
Target Species | Mouse |
For Use With (Application) | Quant |
Gene ID (Entrez) | 13019 |
Quantity | 1 Kit |
Storage Requirements | Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction