Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTGF/CCN2 Antibody (MM0038-6B13) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | CTGF/CCN2 |
---|---|
Clone | MM0038-6B13 |
Concentration | LYOPH |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry, ELISA, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017996
![]() |
Novus Biologicals
NB110610040.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11061004
![]() |
Novus Biologicals
NB11061004 |
0.1 mg | N/A | N/A | N/A | ||||
Description
CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CTGF/CCN2 | |
LYOPH | |
Immunohistochemistry, ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
P29279 | |
1490 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0038-6B13 | |
ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
IgG1 | |
Protein G purified | |
Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title