Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTGF/CCN2 Mouse anti-Human, Clone: 180000000, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | CTGF/CCN2 |
---|---|
Clone | 180000000 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6023550UL
![]() |
Novus Biologicals
NBP26023550UL |
50 μL |
Each for $340.50
|
|
|||||
NB60235100U
![]() |
Novus Biologicals
NBP260235100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CTGF/CCN2 | |
1 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
1490 | |
Recombinant human CTGF (27-349 aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
180000000 | |
Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
CTGF | |
IgG2a κ | |
Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title