Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542811
Description
Peptide sequence: ESNPWVEKVI YPGLPSHPQH ELVKRQCTGC TGMVTFYIKG TLQHAEIFLK Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 93%.
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
CTH | |
Polyclonal | |
Unconjugated | |
CTH | |
0610010I13Rik; AI314617; BC019483; CSE; Cth; cth protein; CTL target antigen; Cys3; cystathionase; cystathionase (cystathionine gamma-lyase); cystathionase isoform 1; cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; cysteine-protein sulfhydrase; EGK_00877; fb58d08; gamma-cystathionase; H; homoserine deaminase; homoserine dehydratase; LOC102117863; MGC9471; PRB-RA; probasin-related antigen; QccE-22305; synthase; Unknown (protein for MGC:85785); wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785 | |
Rabbit | |
Protein A | |
RUO | |
1491 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P32929 | |
CTH | |
Synthetic peptide directed towards the C-terminal of human CTH (aa 281-330). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction