Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTHRC1 Antibody (055), DyLight 405 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289945V
Description
CTHRC1 Monoclonal antibody specifically detects CTHRC1 in Human samples. It is validated for ELISA, ELISASpecifications
CTHRC1 | |
Monoclonal | |
DyLight 405 | |
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CTHRC1 (NP_612464.1; Met1-Lys243). | |
0.1 mL | |
Cancer | |
115908 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
055 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction