Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTHRC1 Antibody (055), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289945MFV610
Description
CTHRC1 Monoclonal antibody specifically detects CTHRC1 in Human samples. It is validated for ELISASpecifications
CTHRC1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
055 | |
Sandwich ELISA Capture | |
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
Rabbit immunized with purified, recombinant Human CTHRC1 (Accession#: NP_612464.1, Met1-Lys243) | |
0.1 mL | |
Cancer | |
115908 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction