Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTHRC1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298449MFV610
Description
CTHRC1 Polyclonal antibody specifically detects CTHRC1 in Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
CTHRC1 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
Produced in rabbits immunized with purified, recombinant Rat CTHRC1 (Accession#: NP_001258229.1; Met1-Lys230). | |
0.1 mL | |
Cancer | |
115908 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction