Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTHRC1 Rabbit anti-Human, Clone: 55, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | CTHRC1 |
---|---|
Clone | 55 |
Dilution | Sandwich ELISA Capture 1:250-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
CTHRC1 Monoclonal specifically detects CTHRC1 in Human samples. It is validated for Sandwich ELISA Capture.Specifications
CTHRC1 | |
Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
115908 | |
IgG | |
Protein A purified |
55 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
Recombinant Human CTHRC1 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title