Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CtIP Antibody (19E8), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

Supplier:  Novus Biologicals NBP141205

Catalog No. NBP141205


Only null left
Add to Cart

Description

Description

CtIP Monoclonal specifically detects CtIP in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay.
Specifications

Specifications

CtIP
Monoclonal
1.3mg/mL
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP)
RBBP8
Human CtIP
Immunogen affinity purified
RUO
5932
Human, Mouse
Purified
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay
1900000000
Unconjugated
CtBP-interacting protein, CtIP, DNA endonuclease RBBP8, EC 3.1, RBBP-8, retinoblastoma binding protein 8, Retinoblastoma-binding protein 8, Retinoblastoma-interacting protein and myosin-like, RIMSAE2, Sporulation in the absence of SPO11 protein 2 homolog
Mouse
125 kDa
100 μL
Primary
This antibody will recognize human CtIP.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only