Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CtIP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501557742
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7742 Supplier Bethyl Laboratories, Inc Supplier No. A300487A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CtIP, a 125 kD protein, was originally found interacting with a transcription repressor, CtBP, through the PLDLS motif (CtBP-Interacting Protein) thus suggested a role in transcription. Studies have shown that CtIP also interacts with BRCA1 protein through the c-terminus BRCT domains also suggested that CtIP is a potential tumor suppressor. This CtIP-BRCA1 interaction can be disruppted by DNA damaging agents including UV or gamma-irradiation. Li et al (Nature 406, 210 - 215 (2000)) have shown that ATM phosphorylates CtIP at serine residues 664 and 745, and mutation of these sites abrogates the dissociation of BRCA1 from CtIP, resulting in persistent repression of BRCA1-dependent induction of GADD45 upon ionizing radiation.

Specifications

Antigen CtIP
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene RBBP8
Gene Accession No. Q99708
Gene Alias CTIP, RIM
Gene Symbols RBBP8
Host Species Rabbit
Immunogen Between 525 and 575
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5932
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.