Learn More
CtIP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300487AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
CtIP, a 125 kD protein, was originally found interacting with a transcription repressor, CtBP, through the PLDLS motif (CtBP-Interacting Protein) thus suggested a role in transcription. Studies have shown that CtIP also interacts with BRCA1 protein through the c-terminus BRCT domains also suggested that CtIP is a potential tumor suppressor. This CtIP-BRCA1 interaction can be disruppted by DNA damaging agents including UV or gamma-irradiation. Li et al (Nature 406, 210 - 215 (2000)) have shown that ATM phosphorylates CtIP at serine residues 664 and 745, and mutation of these sites abrogates the dissociation of BRCA1 from CtIP, resulting in persistent repression of BRCA1-dependent induction of GADD45 upon ionizing radiation.Specifications
CtIP | |
Polyclonal | |
Unconjugated | |
RBBP8 | |
CTIP, RIM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5932 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q99708 | |
RBBP8 | |
Between 525 and 575 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.