Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Antibody (037), DyLight 650

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290616C
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Mouse samples. It is validated for ELISA.Specifications
CTLA-4 | |
Monoclonal | |
DyLight 650 | |
CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CTLA-4 (NP_033973.2; Met1-Phe162). | |
0.1 mL | |
Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
1493 | |
Mouse | |
Purified |
ELISA | |
37 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CTLA4 Mouse CD28 Mouse ICOS Cynomolgus CTLA4 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction