Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Antibody (2188A), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB3252MFV500
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| CTLA-4 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 1493 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry | |
| 2188A | |
| Flow Cytometry, Immunocytochemistry | |
| CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CTLA-4, Ala37-Phe162, Accession # P16410 | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction