Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Antibody (63828), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB434Z
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOFSpecifications
CTLA-4 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
63828 | |
Western Blot, Flow Cytometry, Neutralization, CyTOF-ready | |
CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Mouse myeloma cell line NS0-derived recombinant mouse CTLA-4, Ala36-Phe161, Accession # XP_001479180 | |
0.1 mL | |
Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
1493 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction