Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Antibody (BNI3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB100648490.1MG
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CTLA-4 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry Neat-1:10, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 5 ug/ml-50 ug/ml | |
P16410 | |
CTLA4 | |
Human CTLA-4/human IgG heavy chain fusion protein | |
0.1 mg | |
Signal Transduction | |
1493 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
BNI3 | |
Unconjugated | |
PBS, 0.5% Bovine Serum Albumin with 0.09% Sodium Azide | |
CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
NB100-64849 recognizes the human CD152 cell surface antigen, also known as CTLA-4 (cytotoxic T-lymphocyte-associated antigen 4). CD152 is a 45kD glycoprotein, expressed normally as a disulphide-linked homodimer. It is expressed transiently on the surface of T cells following activation (peaking at 3 days), but is often present with the cytoplasm. CD152 is a ligand for CD80 and CD86, and is important in cellular interactions during the immune response. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction