Learn More
Invitrogen™ CTLA-4 (CD152) Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
$459.00 - $1159.00
Specifications
Antigen | CTLA-4 (CD152) Humanized Biosimilar |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
Classification | Recombinant Monoclonal |
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Antitope patent anti-CTLA4.
CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity than CD28. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhbits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.Specifications
CTLA-4 (CD152) Humanized Biosimilar | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Lyophilized | |
IgG1 | |
Human | |
Primary | |
Protein A |
1 mg/mL | |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
Unconjugated | |
Human | |
RUO | |
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.