Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA4 Rabbit anti-Human, Mouse, Unconjugated, Clone: ADCD-3, Abnova™

Catalog No. 89339766 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-766 Supplier Abnova Corporation Supplier No. MAB22028
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CTLA4 Monoclonal antibody specifically detects CTLA4 in Human, Mouse samples. It is validated for ICC, WB-Ce, IF.

Specifications

Antigen CTLA4
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Monoclonal
Clone ADCD-3
Conjugate Unconjugated
Description cytotoxic T-lymphocyte-associated protein 4
Gene CTLA4
Gene Alias CD152, CELIAC3, CTLA-4, GSE, IDDM12
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CTLA4
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1493
Target Species Human, Mouse
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.