Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTNNA1 Mouse anti-Human, Clone:8B6C1, Abnova™

Catalog No. 89333564 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-564 Supplier Abnova Corporation Supplier No. MAB17307
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CTNNA1.

Specifications

Antigen CTNNA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8B6C1
Conjugate Unconjugated
Description catenin (cadherin-associated protein), alpha 1, 102kDa
Dilution ELISA (1:10000)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CTNNA1
Gene Alias CAP102, FLJ36832
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 371-574 of of human CTNNA1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 1495
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.