Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ctnna1 Mouse anti-Human, Mouse, Clone: alpha-CAT-7A4, Abnova™

Catalog No. 89035391 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-391 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-391 Supplier Abnova Corporation Supplier No. MAB13984

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against synthetic peptide of mouse Ctnna1.

  • High specificity
  • High reactivity

Specifications

Antigen Ctnna1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone alpha-CAT-7A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of mouse Ctnna1.
Dilution ELISA Flow Cytometry (3-5 ug/106 cells) Immunocytochemistry (2 ug/mL) Immunofluorescence (2 ug/mL) Immunohistochemistry Immunoprecipitation Western Blot (2 ug/mL) The optimal working dilution should be determined by the end
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Gene Accession No. P26231
Gene Alias 2010010M04Rik/AA517462/AI988031/Catna1
Gene Symbols Ctnna1
Host Species Mouse
Immunogen A synthetic peptide corresponding to the C-terminus of mouse Ctnna1.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12385
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.