Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTNNB1 Mouse anti-Chicken, Human, Mouse, Clone: CAT-5H10, Abnova™

Catalog No. 89035407 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-407 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-407 Supplier Abnova Corporation Supplier No. MAB14002

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant chicken CTNNB1.

  • High specificity
  • High reactivity

Specifications

Antigen CTNNB1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CAT-5H10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant chicken CTNNB1.
Dilution Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-1:100) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 0.09% sodium azide).
Gene Alias CHBCAT
Gene Symbols CTNNB1
Host Species Mouse
Immunogen Recombinant MBP fusion protein corresponding to a 100 amino acid fragment of the C-terminus of chicken CTNNB1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 395964
Target Species Chicken, Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.