Learn More
CTNNB1 Mouse anti-Human, Mouse, Rat, Clone: CTNNB1/1507, Abnova™
Shop All Abnova Corporation ProductsDescription
Mouse monoclonal antibody raised against full length recombinant human CTNNB1.
- High specificity
- High reactivity
Specifications
Specifications
| Antigen | CTNNB1 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | CTNNB1/1507 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against full length recombinant human CTNNB1. |
| Dilution | Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by |
| Formulation | In 10 mM PBS (0.05% BSA, 0.05% sodium azide). |
| Gene Accession No. | P35222 |
| Gene Alias | CTNNB/DKFZp686D02253/FLJ25606/FLJ37923 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.