Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTNNBIP1 Recombinant Rabbit Monoclonal Antibody (JE58-47)
GREENER_CHOICE

Catalog No. PIMA544879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544879 Supplier Invitrogen™ Supplier No. MA544879
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CTNNBIP1 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene binds CTNNB1 and prevents interaction between CTNNB1 and TCF family members. The encoded protein is a negative regulator of the Wnt signaling pathway. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTNNBIP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE58-47
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CTNNBIP1
Gene Accession No. Q9NSA3
Gene Alias 1110008O09Rik; 2310001I19Rik; AW457332; beta-catenin-interacting protein 1; beta-catenin-interacting protein ICAT; catenin; catenin beta interacting protein 1; catenin, beta interacting protein 1; catenin, beta-interacting protein 1; Catnbip1; Ctnnbip1; ICAT; inhibitor of beta-catenin and Tcf-4
Gene Symbols CTNNBIP1
Host Species Rabbit
Immunogen Synthetic peptide within human CTNNBIP1 aa 1-50/81.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.