Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTNNBL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521112
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521112 Supplier Invitrogen™ Supplier No. PA521112
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-21112 can be used with blocking peptide PEP-1226.

The protein encoded by this gene is a component of the pre-mRNA-processing factor 19-cell division cycle 5-like (PRP19-CDC5L) protein complex, which activates pre-mRNA splicing and is an integral part of the spliceosome. The encoded protein is also a nuclear localization sequence binding protein, and binds to activation-induced deaminase and is important for antibody diversification. This gene may also be associated with the development of obesity. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been defined on the X chromosome. Diseases associated with CTNNBL1 include Immunodeficiency 99 With Hypogammaglobulinemia And Autoimmune Cytopenias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTNNBL1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CTNNBL1
Gene Accession No. Q4V8K2, Q8WYA6, Q9CWL8
Gene Alias 5730471K09Rik; beta catenin-like 1; beta-catenin-like protein 1; C20orf33; catenin beta like 1; catenin, beta like 1; CTNNBL1; dJ633O20.1; NAP; nuclear associated protein; nuclear protein NAP; nuclear-associated protein; NYD-SP19; P14L; PP8304; RP5-1118M15.1; Testis development protein NYD-SP19
Gene Symbols CTNNBL1
Host Species Rabbit
Immunogen A 20 amino acid peptide near the carboxy terminus of human CTTNBL1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296320, 56259, 66642
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.