Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTNNBL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987905
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7905 Supplier Bioss Supplier No. BS7658R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Beta-catenin-like protein 1 (CTNNBL1) contains an acidic domain, a putative bipartite nuclear localization signal, a nuclear export signal, a leucine-isoleucine zipper, and phosphorylation motifs, as well as Armadillo/beta-catenin-like repeats. Transient expression of CTNNBL1 resulted in translocation to the nucleus and apoptosis, suggesting it may be involved in the apoptotic pathway. CTNNBL1 interacts with the Prp19 complex of the spliceosome and the Ig class switching enzyme activation-induced deaminase (AID) and had been suggested to play a role in antibody-diversification and class switching, but recent studies have shown CTNNBL1 to be dispensable for Ig class switch recombination. Other studies have identified CTTNBL1 as a novel gene for obesity.

Specifications

Antigen CTNNBL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CTNNBL1
Gene Accession No. Q8WYA6
Gene Alias 5730471K09Rik; beta catenin-like 1; Beta-catenin-like protein 1; C20orf33; catenin beta like 1; catenin, beta like 1; CTNNBL1; dJ633O20.1; NAP; nuclear associated protein; nuclear protein NAP; Nuclear-associated protein; NYD-SP19; P14L; PP8304; RP5-1118M15.1; Testis development protein NYD-SP19
Gene Symbols CTNNBL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CTNNBL1/Beta catenin like 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56259
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.