Learn More
CTNND1 Mouse anti-Human, Clone: OTI2F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800905

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.Specifications
CTNND1 | |
Monoclonal | |
Unconjugated | |
CTNND1 | |
BCDS2, CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN) | |
Mouse | |
Affinity Chromatography | |
RUO | |
1500 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60716 | |
CTNND1 | |
Full length human recombit protein of human CTNND1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.