Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CTPI 2
GREENER_CHOICESDP

Catalog No. 779410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-941-0 10 mg
77-945-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-941-0 Supplier Tocris Bioscience™ Supplier No. 7794/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective mitochondrial citrate transporter (SLC25A1) inhibitor

CTPI 2 is a selective mitochondrial citrate transporter (SLC25A1) inhibitor with a KD of 3.5 μM. CTPI 2 inhibits the proliferation of H1299 cells in a SLC25A1-dependent manner. It also re-sensitizes T1, T2, and T4 cells to Cisplatin (Cat. No. 2251). CTPI 2 inhibits tumor growth in in vivo models of non-small cell lung cancer (NSCLC). In high-fat diet fed mice CTPI 2 regulates glycolysis, prevents steatohepatitis and normalizes glucose tolerance and insulin sensitivity. CTPI 2 influences the inflammatory pathway by inhibiting IL-6 and TNFα production and M1 macrophage polarization.

Specifications

Chemical Name or Material 2-[[(4-Chloro-3-nitrophenyl)sulfonyl]amino]benzoic acid
CAS 68003-38-3
Quantity 10 mg
Target Other Ion Pump/Transporter Inhibitors
Molecular Formula C13H9ClN2O6S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.