Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113344
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113344 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113344 Supplier Invitrogen™ Supplier No. PA5113344
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTPS2 Polyclonal Antibody for ELISA, IHC (P), IP, Western Blot

The protein encoded by this gene catalyzes the formation of CTP from UTP with the concomitant deamination of glutamine to glutamate. This protein is the rate-limiting enzyme in the synthesis of cytosine nucleotides, which play an important role in various metabolic processes and provide the precursors necessary for the synthesis of RNA and DNA. Cancer cells that exhibit increased cell proliferation also exhibit an increased activity of this encoded protein. Thus, this protein is an attractive target for selective chemotherapy. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTPS2
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ctps2
Gene Accession No. Q9NRF8
Gene Alias A830031M15Rik; AI326475; CTP synthase 2; CTP synthase II; CTP synthetase 2; CTP synthetase homolog; CTP synthetase type 2; CTPS2; CTPsH; cytidine 5'-triphosphate synthase 2; cytidine 5'-triphosphate synthetase 2; cytidine triphosphate synthase II; UTP-ammonia ligase 2; UTP--ammonia ligase 2
Gene Symbols Ctps2
Host Species Rabbit
Immunogen Recombinant Human CTP synthase 2 protein (62-315AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56474
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.