Learn More
Invitrogen™ CTPS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114194
Description
Immunogen sequence: AKRENFCNIH VSLVPQLSAT GEQKTKPTQN SVRALRGLGL SPDLIVCRSS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The protein encoded by this gene catalyzes the formation of CTP from UTP with the concomitant deamination of glutamine to glutamate. This protein is the rate-limiting enzyme in the synthesis of cytosine nucleotides, which play an important role in various metabolic processes and provide the precursors necessary for the synthesis of RNA and DNA. Cancer cells that exhibit increased cell proliferation also exhibit an increased activity of this encoded protein. Thus, this protein is an attractive target for selective chemotherapy. Alternative splicing results in multiple transcript variants.
Specifications
CTPS2 | |
Polyclonal | |
Unconjugated | |
Ctps2 | |
A830031M15Rik; AI326475; CTP synthase 2; CTP synthase II; CTP synthetase 2; CTP synthetase homolog; CTP synthetase type 2; CTPS2; CTPsH; cytidine 5'-triphosphate synthase 2; cytidine 5'-triphosphate synthetase 2; cytidine triphosphate synthase II; UTP-ammonia ligase 2; UTP--ammonia ligase 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56474 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NRF8 | |
Ctps2 | |
Immunogen sequence: AKRENFCNIH VSLVPQLSAT GEQKTKPTQN SVRALRGLGL SPDLIVCRSS. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.