Learn More
Invitrogen™ CTR1 Recombinant Rabbit Monoclonal Antibody (4C10C5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542624
Description
Positive test controls include: 293T, HepG2, A-549, Mouse liver, Mouse kidney, Rat liver. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: MDHSHHMGMS YMDSNSTMQP SHHHPTTSAS HSHGGGDSSM MMMPMTFYFG FKNVELLFSG LVINTAGEMA GAFVAVFLLA MFYEGLKIAR ESLLRKSQVS.
The trace metal copper (Cu) plays a crucial role in mammalian cells as a cofactor for many enzymes. Cu-related genetic diseases, such as Menkes disease and Wilson disease, arise from a lack of Cu homeostasis in mammalian cells. CTR1 is a high-affinity copper-uptake protein. The C-terminal domain is similar to the Raf family of protein kinases, but it's first two-thirds encodes a novel protein domain. CTR1 provides the primary avenue for copper uptake in mammalian cells, thereby, affecting Cu homeostasis and embryonic development.
Specifications
CTR1 | |
Recombinant Monoclonal | |
0.25 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15431, Q8K211, Q9JK41 | |
SLC31A1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CTR1/SLC31A1 (NP_0018501). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
4C10C5 | |
Unconjugated | |
SLC31A1 | |
4930445G01Rik; AI787263; AU016967; copper transport 1 homolog; copper transporter 1; Copper uptake transporter 1; COPT1; ctr1; ctr-1; hCTR1; high affinity copper uptake protein; high affinity copper uptake protein 1; high-affinity copper uptake protein; liver regeneration-related protein LRRGT00200; LRRGT00200; rCTR1; SLC31A1; slc31a1.L; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporter), member 1 L homeolog; solute carrier family 31 (copper transporters), member 1; solute carrier family 31 member 1; solute carrier family 31, member 1; Xctr1; XELAEV_18038194mg; zgc:76839 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1317, 171135, 20529 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.