Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTR2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105199PE
Description
CTR2 Polyclonal antibody specifically detects CTR2 in Human, Mouse samples. It is validated for Western Blot, Western BlotSpecifications
CTR2 | |
Polyclonal | |
Western Blot, Simple Western | |
Copper transporter 2, COPT2SLC13A2, CTR2Solute carrier family 31 member 2, hCTR2probable low affinity copper uptake protein 2, solute carrier family 31 (copper transporters), member 2 | |
Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). [Swiss-Prot# O15432] | |
0.1 mL | |
Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
1318 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction