Learn More
Invitrogen™ CTR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522961
Description
This CTR2 antibody is useful for Western blot, where a band is seen at ~71 kDa. The theoretical molecular weight of CTR2 is 15 kDa. The difference between the expected size and the observed size in Western blot is believed to be due to a multimeric form of CTR2.Suggested positive control: MEF whole cell lysate.
Copper is a trace element that plays an essential role for organisms that utilize oxygen during respiration. CTR2 is a membrane protein that is a member of the SLC31A transporter family. Although the function of CTR2 is still poorly understood in mammalian cells, it is expected to be involved in low-affinity copper uptake. It is believed to be confined to lysosomes which stimulate copper delivery to the the cytosol of human cells at high concentrations.
Specifications
CTR2 | |
Polyclonal | |
Unconjugated | |
SLC31A2 | |
AI604396; Copper transporter 2; COPT2; CTR2; hCTR2; probable low affinity copper uptake protein 2; RP11-9H12.2; SLC31A2; solute carrier family 31 (copper transporter), member 2; solute carrier family 31 (copper transporters), member 2; solute carrier family 31 member 2; solute carrier family 31, member 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1318, 20530 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.93 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
O15432, Q9CPU9 | |
SLC31A2 | |
Synthetic peptide made to an internal portion of the human protein (within residues 50-100). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.