Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTR9 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297398MFV450
Description
CTR9 Polyclonal antibody specifically detects CTR9 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
CTR9 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
Ctr9, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), KIAA0155SH2 domain-binding protein 1, p150TSP, RNA polymerase-associated protein CTR9 homolog, SH2 domain binding protein 1 (tetratricopeptide repeat containing), SH2BP1p150, TPR-containing, SH2-binding phosphoprotein, TSBP | |
Produced in rabbits immunized with E. coli-derived Human CTR9 fragment. | |
0.1 mL | |
Chromatin Research | |
9646 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction