Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTR9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549401
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549401 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549401 Supplier Invitrogen™ Supplier No. PA549401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTR9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTR9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Ctr9
Gene Accession No. Q62018, Q6PD62
Gene Alias AA409336; Ctr9; CTR9 homolog, Paf1/RNA polymerase II complex component; CTR9, Paf1/RNA polymerase II complex component; Ctr9, Paf1/RNA polymerase II complex component, homolog; Ctr9, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae); Kiaa0155; mKIAA0155; p150; p150TSP; RNA polymerase-associated protein CTR9 homolog; SH2 domain binding protein 1 (tetratricopeptide repeat containing); SH2 domain-binding protein 1; SH2BP1; Tetratricopeptide repeat-containing, SH2-binding phosphoprotein of 150 kDa; TPR-containing, SH2-binding phosphoprotein; TPR-containing, SH2-binding phosphoprotein of 150 kDa; TSBP; Tsp
Gene Symbols Ctr9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1022-1056 amino acids from the C-terminal region of Mouse Ctr9.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22083, 9646
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.