Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549154
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549154 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549154 Supplier Invitrogen™ Supplier No. PA549154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTRB1 Polyclonal Antibody for Western Blot

Preferential cleavage: Tyr- -Xaa, Trp- -Xaa, Phe- -Xaa, Leu- -Xaa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CTRB1
Gene Accession No. P17538
Gene Alias 2200008D09Rik; AI504462; Chymotrypsin B; Chymotrypsin B chain A; Chymotrypsin B chain B; Chymotrypsin B chain C; chymotrypsin B1; chymotrypsinogen B; chymotrypsinogen B1; Ctrb; Ctrb1; Prt-2
Gene Symbols CTRB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 145-171 amino acids from the Central region of human CTRB1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.