Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113588 Supplier Invitrogen™ Supplier No. PA5113588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKNPKFSILT VNNDITLLKL ATPARFSQTV SAVCLPSADD DFPAGTLCAT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%.

Preferential cleavage: Tyr- -Xaa, Trp- -Xaa, Phe- -Xaa, Leu- -Xaa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRB1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CTRB1
Gene Accession No. P17538
Gene Alias 2200008D09Rik; AI504462; Chymotrypsin B; Chymotrypsin B chain A; Chymotrypsin B chain B; Chymotrypsin B chain C; chymotrypsin B1; chymotrypsinogen B; chymotrypsinogen B1; Ctrb; Ctrb1; Prt-2
Gene Symbols CTRB1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CTRB1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1504
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.